accessibility menu, dialog, popup

UserName

CAS RN 68077-26-9

IUPAC Name: 7-chloro-1-ethyl-6-fluoro-4-oxo-1,4-dihydroquinoline-3-carboxylate
Synonyms: 7-chloro-1-ethyl-6-fluoro-4-oxoquinoline-3-carboxylate
Molecular Weight (g/mol): 268.65
Molecular Formula: C12H8ClFNO3
InChi Key: WNNSMMJBBOPPOT-UHFFFAOYSA-M
SMILES: CCN1C=C(C([O-])=O)C(=O)C2=CC(F)=C(Cl)C=C12
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results