missing translation for 'onlineSavingsMsg'
Learn More

7-Chloro-1-cyclopropyl-6-fluoro-1,4-dihydro-4-oxo-1,8-naphthyridine-3-carboxylic Acid 98.0+%, TCI America™

Catalog No. C26501G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C26501G 1 g
C26505G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C26501G Supplier TCI America Supplier No. C26501G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 100361-18-0
Molecular Formula C12H8ClFN2O3
Molecular Weight (g/mol) 282.655
MDL Number MFCD08458865
InChI Key OXNZWNNMJBOZQO-UHFFFAOYSA-N
PubChem CID 11055142
IUPAC Name 7-chloro-1-cyclopropyl-6-fluoro-4-oxo-1,8-naphthyridine-3-carboxylic acid
SMILES C1CC1N2C=C(C(=O)C3=CC(=C(N=C32)Cl)F)C(=O)O

Specifications

Melting Point 224°C
Color White-Yellow
Quantity 1 g
Formula Weight 282.66
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 7-Chloro-1-cyclopropyl-6-fluoro-1,4-dihydro-4-oxo-1,8-naphthyridine-3-carboxylic Acid