missing translation for 'onlineSavingsMsg'
Learn More

7-Chloro-1,2,3,4-tetrahydro-5H-1-benzazepin-5-one 98.0+%, TCI America™

Catalog No. C28965G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C28965G 5 g
C28961G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C28965G Supplier TCI America Supplier No. C28965G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 160129-45-3
Molecular Formula C10H10ClNO
Molecular Weight (g/mol) 195.65
MDL Number MFCD06738694
InChI Key AHESNFIUAHTYGS-UHFFFAOYSA-N
Synonym 7-Chloro-3,4-dihydro-1H-benzo[b]azepin-5(2H)-one
PubChem CID 22903483
IUPAC Name 7-chloro-2,3,4,5-tetrahydro-1H-1-benzazepin-5-one
SMILES ClC1=CC2=C(NCCCC2=O)C=C1

Specifications

Melting Point 105°C
Quantity 5 g
Formula Weight 195.65
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 7-Chloro-1,2,3,4-tetrahydro-5H-1-benzazepin-5-one