accessibility menu, dialog, popup

UserName

CAS RN 160129-45-3

IUPAC Name: 7-chloro-2,3,4,5-tetrahydro-1H-1-benzazepin-5-one
Synonyms: 7-chloro-1,2,3,4-tetrahydro-1-benzazepin-5-one
Molecular Weight (g/mol): 195.65
Molecular Formula: C10H10ClNO
InChi Key: AHESNFIUAHTYGS-UHFFFAOYSA-N
SMILES: ClC1=CC2=C(NCCCC2=O)C=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results