missing translation for 'onlineSavingsMsg'
Learn More
Learn More
7-Bromopyrrolo[2,1-f][1,2,4]triazin-4-amine, Thermo Scientific™
$223.32 - $758.23
Chemical Identifiers
| CAS | 937046-98-5 |
|---|---|
| Molecular Formula | C6H5BrN4 |
| Molecular Weight (g/mol) | 213.04 |
| MDL Number | MFCD09033848 |
| InChI Key | YGCJBESZJIGDMP-UHFFFAOYSA-N |
| IUPAC Name | 7-bromopyrrolo[2,1-f][1,2,4]triazin-4-amine |
| SMILES | NC1=NC=NN2C(Br)=CC=C12 |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AC468700010
|
Thermo Scientific Chemicals
468700010 |
1 g |
Each for $223.32
|
|
|||||
|
AC468700050
|
Thermo Scientific Chemicals
468700050 |
5 g |
Each for $758.23
|
|
|||||
Chemical Identifiers
| 937046-98-5 | |
| 213.04 | |
| YGCJBESZJIGDMP-UHFFFAOYSA-N | |
| NC1=NC=NN2C(Br)=CC=C12 |
| C6H5BrN4 | |
| MFCD09033848 | |
| 7-bromopyrrolo[2,1-f][1,2,4]triazin-4-amine |
Specifications
| 937046-98-5 | |
| Glass Bottle | |
| MFCD09033848 | |
| YGCJBESZJIGDMP-UHFFFAOYSA-N | |
| 7-bromopyrrolo[2,1-f][1,2,4]triazin-4-amine | |
| 213.04 | |
| 7-Bromopyrrolo[2,1-f][1,2,4]triazin-4-amine |
| Conforms to structure | |
| C6H5BrN4 | |
| 1 g | |
| NC1=NC=NN2C(Br)=CC=C12 | |
| 213.04 | |
| Powder |
Safety and Handling
GHS07: Exclamation mark
ShelfLife : 5 years
Recommended Storage : Refrigerator +4°C