accessibility menu, dialog, popup

UserName

NNNBrNH2

CAS RN 937046-98-5

IUPAC Name: 7-bromopyrrolo[2,1-f][1,2,4]triazin-4-amine
Synonyms: 7-bromopyrrolo[2,1-f][1,2,4]triazin-4-amine
Molecular Weight (g/mol): 213.04
Molecular Formula: C6H5BrN4
InChi Key: YGCJBESZJIGDMP-UHFFFAOYSA-N
SMILES: NC1=NC=NN2C(Br)=CC=C12

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results