missing translation for 'onlineSavingsMsg'
Learn More

7-Bromopyrrolo[2,1-f][1,2,4]triazin-4-amine, Thermo Scientific™

Catalog No. AC468700010
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC468700010 1 g
AC468700050 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC468700010 Supplier Thermo Scientific Chemicals Supplier No. 468700010

Chemical Identifiers

CAS 937046-98-5
Molecular Formula C6H5BrN4
Molecular Weight (g/mol) 213.04
MDL Number MFCD09033848
InChI Key YGCJBESZJIGDMP-UHFFFAOYSA-N
IUPAC Name 7-bromopyrrolo[2,1-f][1,2,4]triazin-4-amine
SMILES NC1=NC=NN2C(Br)=CC=C12

Specifications

Infrared Spectrum Conforms to structure
Packaging Glass Bottle
Quantity 1 g
Formula Weight 213.04
Physical Form Powder
Chemical Name or Material 7-Bromopyrrolo[2,1-f][1,2,4]triazin-4-amine
Hazard Category GHS07: Exclamation mark
ShelfLife 5 years
Recommended Storage Refrigerator +4°C