Learn More
7-Bromoheptanenitrile, 98%
CAS: 20965-27-9 | C7H12BrN | 190.08 g/mol
Supplier: Thermo Scientific Chemicals L0506814
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| 7-Bromoheptanenitrile | |
| 1.266 | |
| >110°C (230°F) | |
| 1.475 | |
| 25 g | |
| 1748070 | |
| HVVQSKCGHAPHMV-UHFFFAOYSA-N | |
| 7-bromoheptanenitrile | |
| 88737 | |
| 98% |
| 20965-27-9 | |
| 114°C to 115°C (4 mmHg) | |
| C7H12BrN | |
| MFCD00001982 | |
| UN3276 | |
| 7-bromoheptanonitrile, heptanenitrile, 7-bromo, 6-cyanohexyl bromide, 1-bromo-6-cyanohexane, 6-bromohexylcyanide, 6-bromohexyl cyanide, 7-bromo-heptanonitrile, 7-bromoheptan enitrile, 7-bromanylheptanenitrile, acmc-1cc07 | |
| BrCCCCCCC#N | |
| 190.08 | |
| 190.09 |
Chemical Identifiers
| 20965-27-9 | |
| 190.08 | |
| HVVQSKCGHAPHMV-UHFFFAOYSA-N | |
| 88737 | |
| BrCCCCCCC#N |
| C7H12BrN | |
| MFCD00001982 | |
| 7-bromoheptanonitrile, heptanenitrile, 7-bromo, 6-cyanohexyl bromide, 1-bromo-6-cyanohexane, 6-bromohexylcyanide, 6-bromohexyl cyanide, 7-bromo-heptanonitrile, 7-bromoheptan enitrile, 7-bromanylheptanenitrile, acmc-1cc07 | |
| 7-bromoheptanenitrile |
Safety and Handling
GHS H Statement
H331-H302-H312-H315-H319
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H302+H312+H332-H315-H319-H335
DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, LIQUID, TOXIC, N.O.S.
EINECSNumber : 244-128-7
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only