missing translation for 'onlineSavingsMsg'
Learn More

7-Bromoheptanenitrile, 98%

Catalog No. AAL0506806
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0506806 5 g
AAL0506814 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0506806 Supplier Thermo Scientific Chemicals Supplier No. L0506806
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 20965-27-9 | C7H12BrN | 190.08 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 20965-27-9
Molecular Formula C7H12BrN
Molecular Weight (g/mol) 190.08
MDL Number MFCD00001982
InChI Key HVVQSKCGHAPHMV-UHFFFAOYSA-N
Synonym 7-bromoheptanonitrile, heptanenitrile, 7-bromo, 6-cyanohexyl bromide, 1-bromo-6-cyanohexane, 6-bromohexylcyanide, 6-bromohexyl cyanide, 7-bromo-heptanonitrile, 7-bromoheptan enitrile, 7-bromanylheptanenitrile, acmc-1cc07
PubChem CID 88737
IUPAC Name 7-bromoheptanenitrile
SMILES BrCCCCCCC#N

Specifications

Density 1.266
Boiling Point 114°C to 115°C (4 mmHg)
Flash Point >110°C (230°F)
Refractive Index 1.475
Quantity 5 g
UN Number UN3276
Beilstein 1748070
Formula Weight 190.09
Percent Purity 98%
Chemical Name or Material 7-Bromoheptanenitrile
Hazard Category H302+H312+H332-H315-H319-H335
Hazard Statement GHS H Statement
H331-H302-H312-H315-H319
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, LIQUID, TOXIC, N.O.S.
EINECSNumber 244-128-7
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only