missing translation for 'onlineSavingsMsg'
Learn More

7-Bromobenz[a]anthracene 98.0+%, TCI America™

Catalog No. p-7119148
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B16031G 1 g
B16035G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B16031G Supplier TCI America Supplier No. B16031G
Only null left

Chemical Identifiers

CAS 32795-84-9
Molecular Formula C18H11Br
Molecular Weight (g/mol) 307.19
MDL Number MFCD00029016
InChI Key LGRNWCDRODWMOH-UHFFFAOYSA-N
PubChem CID 72849
IUPAC Name 7-bromobenzo[a]anthracene
SMILES C1=CC=C2C(=C1)C=CC3=C(C4=CC=CC=C4C=C32)Br

Specifications

Melting Point 152°C
Quantity 1 g
Formula Weight 307.19
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 7-Bromobenz[a]anthracene
RTECSNumber CW0180000
TSCA No