missing translation for 'onlineSavingsMsg'
Learn More

7-Bromo-6-chloro-7-deazapurine, 97%, Thermo Scientific Chemicals

Catalog No. AAH5983303
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5983303 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH5983303 Supplier Thermo Scientific Chemicals Supplier No. H5983303
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 22276-95-5 | Molecular Formula: C6H3BrClN3 | Molecular Weight: 232.465 g/mol

It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.

Solubility
Slightly soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.

Chemical Identifiers

CAS 22276-95-5
Molecular Formula C6H3BrClN3
Molecular Weight (g/mol) 232.465
MDL Number MFCD09702029
InChI Key OXLMTRZWMHIZBY-UHFFFAOYSA-N
Synonym 5-bromo-4-chloro-7h-pyrrolo 2,3-d pyrimidine, 7-bromo-6-chloro-7-deazapurine, 4-chloro-5-bromo-7h-pyrrolo 2,3-d pyrimidine, 7h-pyrrolo 2,3-d pyrimidine, 5-bromo-4-chloro, 7-deaza-7-bromo-6-chloropurine, 5-bromo-4-chloropyrrolo 2,3-d pyrimidine, 5-bromo-4-chloro-1h-pyrrolo 2,3-d pyrimidine, 7-bromo-5-chloro-2,4,9-triazabicyclo 4.3.0 nona-2,4,7,10-tetraene, pubchem14950, acmc-209zzo
PubChem CID 5380710
IUPAC Name 5-bromo-4-chloro-7H-pyrrolo[2,3-d]pyrimidine
SMILES C1=C(C2=C(N1)N=CN=C2Cl)Br

Specifications

Melting Point 221°C to 225°C
Quantity 1 g
Solubility Information Slightly soluble in water.
Formula Weight 232.47
Percent Purity 97%
Chemical Name or Material 7-Bromo-6-chloro-7-deazapurine
Hazard Category H301
Hazard Statement GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P264b-P270-P301+P310-P330-P501c
DOTInformation Hazard Class: 6.1; Packaging Group: III
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only