CAS RN 22276-95-5
IUPAC Name:
5-bromo-4-chloro-7H-pyrrolo[2,3-d]pyrimidine
Synonyms:
5-bromo-4-chloro-7H-pyrrolo[2,3-d]pyrimidine
Molecular Weight (g/mol):
232.47
Molecular Formula:
C6H3BrClN3
InChi Key:
OXLMTRZWMHIZBY-UHFFFAOYSA-N
SMILES:
ClC1=C2C(Br)=CNC2=NC=N1
1
–
1
of
1
results
1
Promotions Available
7-Bromo-6-chloro-7-deazapurine, 97%, Thermo Scientific Chemicals
7-Bromo-6-chloro-7-deazapurine, 97%, Thermo Scientific Chemicals
CAS: 22276-95-5 Molecular Formula: C6H3BrClN3 Molecular Weight (g/mol): 232.465 MDL Number: MFCD09702029 InChI Key: OXLMTRZWMHIZBY-UHFFFAOYSA-N Synonym: 5-bromo-4-chloro-7h-pyrrolo 2,3-d pyrimidine,7-bromo-6-chloro-7-deazapurine,4-chloro-5-bromo-7h-pyrrolo 2,3-d pyrimidine,7h-pyrrolo 2,3-d pyrimidine, 5-bromo-4-chloro,7-deaza-7-bromo-6-chloropurine,5-bromo-4-chloropyrrolo 2,3-d pyrimidine,5-bromo-4-chloro-1h-pyrrolo 2,3-d pyrimidine,7-bromo-5-chloro-2,4,9-triazabicyclo 4.3.0 nona-2,4,7,10-tetraene,pubchem14950,acmc-209zzo PubChem CID: 5380710 IUPAC Name: 5-bromo-4-chloro-7H-pyrrolo[2,3-d]pyrimidine SMILES: C1=C(C2=C(N1)N=CN=C2Cl)Br