missing translation for 'onlineSavingsMsg'
Learn More

7-Bromo-5-methylbenzo[b]furan, 97%

Catalog No. AAH34347MD
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH34347MD 250 mg
AAH3434703 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH34347MD Supplier Thermo Scientific Chemicals Supplier No. H34347MD
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 35700-48-2 | C9H7BrO | 211.058 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 35700-48-2
Molecular Formula C9H7BrO
Molecular Weight (g/mol) 211.058
MDL Number MFCD11877828
InChI Key TZUMDTURHSPEAY-UHFFFAOYSA-N
Synonym 7-bromo-5-methylbenzofuran, 7-bromo-5-methylbenzo b furan, 7-bromo-5-methyl-benzofuran, benzofuran, 7-bromo-5-methyl
PubChem CID 21071801
IUPAC Name 7-bromo-5-methyl-1-benzofuran
SMILES CC1=CC(=C2C(=C1)C=CO2)Br

Specifications

Quantity 250 mg
Formula Weight 211.06
Percent Purity 97%
Chemical Name or Material 7-Bromo-5-methylbenzo[b]furan
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only