missing translation for 'onlineSavingsMsg'
Learn More

7-Bromo-3,4-dihydro-2H-pyrido[3,2-b][1,4]oxazine, 97%, Thermo Scientific™

Catalog No. CC60810DA
Change view
Click to view available options
Quantity:
1g
5g
Packaging:
Amber Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
CC60810DA 1g Amber Glass Bottle
CC60810DE 5g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. CC60810DA Supplier Thermo Scientific Chemicals Supplier No. CC60810DA

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 34950-82-8
Molecular Formula C7H7BrN2O
Molecular Weight (g/mol) 215.05
MDL Number MFCD09025906
InChI Key CYISPVTTZWJFEO-UHFFFAOYSA-N
Synonym 7-bromo-3,4-dihydro-2h-pyrido 3,2-b 1,4 oxazine, 7-bromo-3,4-dihydro-2h-pyrido 3,2-b-1,4-oxazine, 7-bromo-2h,3h,4h-pyrido 3,2-b 1,4 oxazine, 7-bromo-2h,3h,4h-pyridino 2,3-e 1,4-oxazine, pubchem18740, acmc-1aimz, abbypharma ap-13-10238, 2h-pyrido 3,2-b-1,4-oxazine,7-bromo-3,4-dihydro
PubChem CID 24229635
IUPAC Name 7-bromo-2H,3H,4H-pyrido[3,2-b][1,4]oxazine
SMILES BrC1=CN=C2NCCOC2=C1

Specifications

Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1g
Formula Weight 215.05
Percent Purity 97%
Chemical Name or Material 7-Bromo-3,4-dihydro-2H-pyrido[3,2-b][1,4]oxazine