accessibility menu, dialog, popup

UserName

ONNHBr

CAS RN 34950-82-8

IUPAC Name: 7-bromo-2H,3H,4H-pyrido[3,2-b][1,4]oxazine
Synonyms: 7-bromo-2H,3H,4H-pyrido[3,2-b][1,4]oxazine
Molecular Weight (g/mol): 215.05
Molecular Formula: C7H7BrN2O
InChi Key: CYISPVTTZWJFEO-UHFFFAOYSA-N
SMILES: BrC1=CN=C2NCCOC2=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (1)