missing translation for 'onlineSavingsMsg'
Learn More

7-Bromo-2H-1,4-benzothiazin-3(4H)-one 98.0+%, TCI America™

Catalog No. p-7123828
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B49021G 1 g
B49025G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B49021G Supplier TCI America Supplier No. B49021G
Only null left

Chemical Identifiers

CAS 90814-91-8
Molecular Formula C8H6BrNOS
Molecular Weight (g/mol) 244.11
MDL Number MFCD02660583
InChI Key MDFPWMKBRDOSGB-UHFFFAOYSA-N
Synonym 7-bromo-2h-1,4-benzothiazin-3 4h-one, 7-bromo-2h-benzo b 1,4 thiazin-3 4h-one, 7-bromo-3,4-dihydro-2h-1,4-benzothiazin-3-one, 7-bromo-2h,4h-benzo e 1,4-thiazin-3-one, 7-bromo-2,4-dihydro-1,4-benzothiazin-3-one, 2h-1,4-benzothiazin-3 4h-one, 7-bromo, 7-bromo-4h-benzo 1,4 thiazin-3-one
PubChem CID 689065
IUPAC Name 7-bromo-3,4-dihydro-2H-1,4-benzothiazin-3-one
SMILES BrC1=CC2=C(NC(=O)CS2)C=C1

Specifications

Melting Point 214°C
Quantity 1 g
Formula Weight 244.11
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 7-Bromo-2H-1,4-benzothiazin-3(4H)-one