accessibility menu, dialog, popup

UserName

ONHSBr

CAS RN 90814-91-8

IUPAC Name: 7-bromo-3,4-dihydro-2H-1,4-benzothiazin-3-one
Synonyms: 7-bromo-2,4-dihydro-1,4-benzothiazin-3-one
Molecular Weight (g/mol): 244.11
Molecular Formula: C8H6BrNOS
InChi Key: MDFPWMKBRDOSGB-UHFFFAOYSA-N
SMILES: BrC1=CC2=C(NC(=O)CS2)C=C1
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)