missing translation for 'onlineSavingsMsg'
Learn More

7-Bromo-2H-1,4-benzothiazin-3(4H)-one, 97%

Catalog No. AAH2756303
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH2756303 1 g
AAH2756306 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH2756303 Supplier Thermo Scientific Chemicals Supplier No. H2756303
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 90814-91-8 | C8H6BrNOS | 244.11 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 90814-91-8
Molecular Formula C8H6BrNOS
Molecular Weight (g/mol) 244.11
MDL Number MFCD02660583
InChI Key MDFPWMKBRDOSGB-UHFFFAOYSA-N
Synonym 7-bromo-2h-1,4-benzothiazin-3 4h-one, 7-bromo-2h-benzo b 1,4 thiazin-3 4h-one, 7-bromo-3,4-dihydro-2h-1,4-benzothiazin-3-one, 7-bromo-2h,4h-benzo e 1,4-thiazin-3-one, 7-bromo-2,4-dihydro-1,4-benzothiazin-3-one, 2h-1,4-benzothiazin-3 4h-one, 7-bromo, 7-bromo-4h-benzo 1,4 thiazin-3-one
PubChem CID 689065
SMILES BrC1=CC2=C(NC(=O)CS2)C=C1

Specifications

Melting Point 210°C to 214°C
Quantity 1 g
IUPAC Name 7-bromo-3,4-dihydro-2H-1,4-benzothiazin-3-one
Formula Weight 244.11
Percent Purity 97%
Chemical Name or Material 7-Bromo-2H-1,4-benzothiazin-3(4H)-one
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only