missing translation for 'onlineSavingsMsg'
Learn More

7-Bromo-1-tetralone 95.0+%, TCI America™

Catalog No. B45451G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B45451G 1 g
B45455G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B45451G Supplier TCI America Supplier No. B45451G

Chemical Identifiers

CAS 32281-97-3
Molecular Formula C10H9BrO
Molecular Weight (g/mol) 225.085
MDL Number MFCD02179287
InChI Key YGVDCGFUUUJCDF-UHFFFAOYSA-N
Synonym 7-bromo-1-tetralone, 7-bromo-3,4-dihydronaphthalen-1 2h-one, 7-bromo-3,4-dihydro-1 2h-naphthalenone, 7-bromotetralone, 7-bromo-alpha-tetralone, 7-bromotetralin-1-one, 7-bromo-1,2,3,4-tetrahydronaphthalen-1-one, 7-bromo-2,3,4-trihydronaphthalen-1-one, 1 2h-naphthalenone, 7-bromo-3,4-dihydro, 7-bromo tetralone
PubChem CID 252731
IUPAC Name 7-bromo-3,4-dihydro-2H-naphthalen-1-one
SMILES C1CC2=C(C=C(C=C2)Br)C(=O)C1

Specifications

Melting Point 76°C
Color White-Yellow
Boiling Point 157°C
Quantity 1 g
Formula Weight 225.09
Percent Purity ≥95.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 7-Bromo-1-tetralone