missing translation for 'onlineSavingsMsg'
Learn More

7-Bromo-1-chloroisoquinoline, 97%, Thermo Scientific Chemicals

Catalog No. AAL1994606
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1994606 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAL1994606 Supplier Thermo Scientific Chemicals Supplier No. L1994606

7-Bromo-1-chloroisoquinoline as an organic chemical synthesis intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
7-Bromo-1-chloroisoquinoline as an organic chemical synthesis intermediate.

Solubility
Insoluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.

Chemical Identifiers

CAS 215453-51-3
Molecular Formula C9H5BrClN
Molecular Weight (g/mol) 242.5
MDL Number MFCD04039320
InChI Key UMSWWSIVPWVJOX-UHFFFAOYSA-N
Synonym 7-bromo-1-chloro-isoquinoline, 1-chloro-7-bromoisoquinoline, isoquinoline, 7-bromo-1-chloro, acmc-209y3w
PubChem CID 6502024
IUPAC Name 7-bromo-1-chloroisoquinoline
SMILES C1=CC(=CC2=C1C=CN=C2Cl)Br

Specifications

Melting Point 122°C to 126°C
Quantity 5 g
Beilstein 8404252
Solubility Information Insoluble in water.
Formula Weight 242.5
Percent Purity 97%
Chemical Name or Material 7-Bromo-1-chloroisoquinoline
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 000-000-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only