CAS RN 215453-51-3
IUPAC Name:
7-bromo-1-chloroisoquinoline
Synonyms:
7-bromo-1-chloroisoquinoline
Molecular Weight (g/mol):
242.5
Molecular Formula:
C9H5BrClN
InChi Key:
UMSWWSIVPWVJOX-UHFFFAOYSA-N
SMILES:
ClC1=NC=CC2=CC=C(Br)C=C12
1
–
1
of
1
results
1
7-Bromo-1-chloroisoquinoline, 97%, Thermo Scientific Chemicals
CAS: 215453-51-3 Molecular Formula: C9H5BrClN Molecular Weight (g/mol): 242.5 MDL Number: MFCD04039320 InChI Key: UMSWWSIVPWVJOX-UHFFFAOYSA-N Synonym: 7-bromo-1-chloro-isoquinoline,1-chloro-7-bromoisoquinoline,isoquinoline, 7-bromo-1-chloro,acmc-209y3w PubChem CID: 6502024 IUPAC Name: 7-bromo-1-chloroisoquinoline SMILES: C1=CC(=CC2=C1C=CN=C2Cl)Br