missing translation for 'onlineSavingsMsg'
Learn More

7-Bromo-1,2,3,4-tetrahydroisoquinoline, 97%

Catalog No. AAB2571203
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2571203 1 g
AAB2571206 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2571203 Supplier Thermo Scientific Chemicals Supplier No. B2571203
Only null left

CAS: 17680-55-6 | C9H10BrN | 212.09 g/mol

7-Bromo-1,2,3,4-tetrahydroisoquinoline is used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
7-Bromo-1,2,3,4-tetrahydroisoquinoline is used as a pharmaceutical intermediate.

Solubility
Insoluble in water.

Notes
Store in a cool, dry place in tightly closed container. Incompatible with oxidizing agents.

Chemical Identifiers

CAS 17680-55-6
Molecular Formula C9H10BrN
Molecular Weight (g/mol) 212.09
MDL Number MFCD06739047
InChI Key OYODEQFZAJVROF-UHFFFAOYSA-N
Synonym 7-bromo-1,2,3,4-tetrahydro-isoquinoline, isoquinoline, 7-bromo-1,2,3,4-tetrahydro, acmc-209uo4, 7-bromotetrahydroisoquinoline
PubChem CID 10729255
IUPAC Name 7-bromo-1,2,3,4-tetrahydroisoquinoline
SMILES C1CNCC2=C1C=CC(=C2)Br

Specifications

Melting Point 32°C to 35°C
Quantity 1 g
Solubility Information Insoluble in water.
Formula Weight 212.09
Percent Purity 97%
Chemical Name or Material 7-Bromo-1,2,3,4-tetrahydroisoquinoline
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only