CAS RN 17680-55-6
IUPAC Name:
7-bromo-1,2,3,4-tetrahydroisoquinoline
Synonyms:
7-bromo-1,2,3,4-tetrahydroisoquinoline
Molecular Weight (g/mol):
212.09
Molecular Formula:
C9H10BrN
InChi Key:
OYODEQFZAJVROF-UHFFFAOYSA-N
SMILES:
BrC1=CC2=C(CCNC2)C=C1
1
–
1
of
1
results
1
Promotions Available
7-Bromo-1,2,3,4-tetrahydroisoquinoline, 97%
7-Bromo-1,2,3,4-tetrahydroisoquinoline, 97%
CAS: 17680-55-6 Molecular Formula: C9H10BrN Molecular Weight (g/mol): 212.09 MDL Number: MFCD06739047 InChI Key: OYODEQFZAJVROF-UHFFFAOYSA-N Synonym: 7-bromo-1,2,3,4-tetrahydro-isoquinoline,isoquinoline, 7-bromo-1,2,3,4-tetrahydro,acmc-209uo4,7-bromotetrahydroisoquinoline PubChem CID: 10729255 IUPAC Name: 7-bromo-1,2,3,4-tetrahydroisoquinoline SMILES: C1CNCC2=C1C=CC(=C2)Br