missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Chemical Identifiers
| CAS | 754214-56-7 |
|---|---|
| Molecular Formula | C13H17BN2O2 |
| Molecular Weight (g/mol) | 244.1 |
| InChI Key | UOXAMYZTYZLSCC-UHFFFAOYSA-N |
| Synonym | 5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-1h-pyrrolo 2,3-b pyridine, 7-azaindole-5-boronic acid pinacol ester, 5-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl-1h-pyrrolo 2,3-b pyridine, 5-tetramethyl-1,3,2-dioxaborolan-2-yl-1h-pyrrolo 2,3-b pyridine, pyrrolo 2,3-b pyridine-5-boronic acid, pinacol ester, pyrrolo 2,3-b pyridin-5-ylboronic acid pinacol ester, zlchem 1246, pubchem16626, acmc-209szp, ksc641k4p |
| PubChem CID | 24208789 |
| IUPAC Name | 5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrrolo[2,3-b]pyridine |
| SMILES | B1(OC(C(O1)(C)C)(C)C)C2=CC3=C(NC=C3)N=C2 |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|
AC455850010
|
N/A | 1g |
N/A
|
N/A | |||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Chemical Identifiers
| 754214-56-7 | |
| 244.1 | |
| 5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-1h-pyrrolo 2,3-b pyridine, 7-azaindole-5-boronic acid pinacol ester, 5-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl-1h-pyrrolo 2,3-b pyridine, 5-tetramethyl-1,3,2-dioxaborolan-2-yl-1h-pyrrolo 2,3-b pyridine, pyrrolo 2,3-b pyridine-5-boronic acid, pinacol ester, pyrrolo 2,3-b pyridin-5-ylboronic acid pinacol ester, zlchem 1246, pubchem16626, acmc-209szp, ksc641k4p | |
| 5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrrolo[2,3-b]pyridine |
| C13H17BN2O2 | |
| UOXAMYZTYZLSCC-UHFFFAOYSA-N | |
| 24208789 | |
| B1(OC(C(O1)(C)C)(C)C)C2=CC3=C(NC=C3)N=C2 |
Specifications
| 754214-56-7 | |
| 96% min. (GC) | |
| C13H17BN2O2 | |
| UOXAMYZTYZLSCC-UHFFFAOYSA-N | |
| 5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrrolo[2,3-b]pyridine | |
| 24208789 | |
| 97% |
| Authentic | |
| Glass Bottle | |
| 5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-1h-pyrrolo 2,3-b pyridine, 7-azaindole-5-boronic acid pinacol ester, 5-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl-1h-pyrrolo 2,3-b pyridine, 5-tetramethyl-1,3,2-dioxaborolan-2-yl-1h-pyrrolo 2,3-b pyridine, pyrrolo 2,3-b pyridine-5-boronic acid, pinacol ester, pyrrolo 2,3-b pyridin-5-ylboronic acid pinacol ester, zlchem 1246, pubchem16626, acmc-209szp, ksc641k4p | |
| B1(OC(C(O1)(C)C)(C)C)C2=CC3=C(NC=C3)N=C2 | |
| 244.1 | |
| 244.1 | |
| 7-Azaindole-5-boronic acid pinacol ester |
Safety and Handling
GHS H Statement Harmful if swallowed. Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation.
GHS P Statement Wear protective gloves/protective clothing/eye protection/face protection. IF ON SKIN: Wash with plenty of soap and water. IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Call a POISON CENTER or doctor/physician if you feel unwell.
GHS Signal Word: Warning
RUO – Research Use Only