missing translation for 'onlineSavingsMsg'
Learn More

(7-Azabenzotriazol-1-yloxy)tris(dimethylamino)phosphonium Hexafluorophosphate 98.0+%, TCI America™

Catalog No. A27825G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A27825G 5 g
A27821G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A27825G Supplier TCI America Supplier No. A27825G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 156311-85-2
Molecular Formula C11H21F6N7OP2
Molecular Weight (g/mol) 443.275
MDL Number MFCD09263289
InChI Key RQBNNDQCKMIUQJ-UHFFFAOYSA-N
Synonym 3h-1,2,3 triazolo 4,5-b pyridin-3-yl oxy tris dimethylamino phosphonium hexafluorophosphate v, 7-azabenzotriazol-1-yloxytris dimethylamino phosphonium hexafluorophosphate, 7-azabenzotriazol-1-yloxytris dimethylamino phosph, 7-aza-1h-benzotriazol-1-yloxytris dimethylamino phosphonium hexafluorophosphate, acmc-20alxg, tris dimethylamino 3h-1,2,3-triazolo 4,5-b pyridin-3-yloxy phosphonium hexafluorophosphate, ksc173q2f, 7-azabenzotriazol-1-yloxytris dimethyamino phosphonium hexafluorosphate, 7-azabenzotriazol-1-yloxy tris dimethylamino phosphonium hexafluorophosphate, 7-aza-1h-benzotriazol-1-yloxytris dimethylamino phosphonium hexafluorophosphate, 98+%
PubChem CID 21894514
IUPAC Name tris(dimethylamino)-(triazolo[4,5-b]pyridin-3-yloxy)phosphanium;hexafluorophosphate
SMILES CN(C)[P+](N(C)C)(N(C)C)ON1C2=C(C=CC=N2)N=N1.F[P-](F)(F)(F)(F)F

Specifications

Melting Point 177°C
Color White
Quantity 5 g
Formula Weight 443.27
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material (7-Azabenzotriazol-1-yloxy)tris(dimethylamino)phosphonium Hexafluorophosphate
TSCA No
Recommended Storage Refrigerator