accessibility menu, dialog, popup

UserName

CAS RN 156311-85-2

IUPAC Name: hexafluoro-λ⁵-phosphanuide; tris(dimethylamino)({3H-[1,2,3]triazolo[4,5-b]pyridin-3-yloxy})phosphanium
Synonyms: tris(dimethylamino)({[1,2,3]triazolo[4,5-b]pyridin-3-yloxy})phosphanium hexafluorophosphate
Molecular Weight (g/mol): 443.28
Molecular Formula: C11H21F6N7OP2
InChi Key: RQBNNDQCKMIUQJ-UHFFFAOYSA-N
SMILES: F[P-](F)(F)(F)(F)F.CN(C)[P+](ON1N=NC2=CC=CN=C12)(N(C)C)N(C)C
Percent Purity 
  • (1)
  • (2)
Quantity 
  • (1)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
  • (2)