missing translation for 'onlineSavingsMsg'
Learn More

7-Aminodesacetoxycephalosporanic Acid, British Pharmacopoeia (BP) Reference Standard, MilliporeSigma™ Supelco™

Catalog No. 111022832
Change view
Click to view available options
Quantity:
20 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111022832 20 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111022832 Supplier Merck Emd Millipore Supplier No. BP011

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets, has been developed and issued under the Authority of the issuing Pharmacopoeia.

Specifications

Chemical Name or Material Pharmacopeia & Metrological Institutes Standards
Molecular Formula C8H10N2O3S
Linear Formula C8H10N2O3S
MDL Number MFCD09750368
Quantity 20 mg
Synonym 7-ADCA
Formula Weight 214.24
Grade British Pharmacopoeia (BP) Reference Standard