missing translation for 'onlineSavingsMsg'
Learn More

7-Aminocephalosporanic Acid 97.0+%, TCI America™

Catalog No. p-7117338
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
25 g
5 g
Catalog No. Quantity
A12665G 5 g
A126625G 25 g
2 options

Catalog No. A12665G

Supplier: TCI America A12665G

Only null left

Chemical Identifiers

CAS 957-68-6
Molecular Formula C10H12N2O5S
Molecular Weight (g/mol) 272.275
MDL Number MFCD00005177
InChI Key HSHGZXNAXBPPDL-HZGVNTEJSA-N
Synonym 7-aminocephalosporanic acid, 7-aca, 7-aminocephalosporinic acid, 7beta-aminocephalosporanic acid, 7-acs, unii-9xi67897rg, 7r-7-aminocephalosporanic acid, 7r-7-aminocephalosporanate, cephalosporanic acid, 7-amino, 6r,7r-3-acetoxymethyl-7-amino-8-oxo-5-thia-1-azabicyclo 4.2.0 oct-2-ene-2-carboxylic acid
PubChem CID 441328
ChEBI CHEBI:2255
IUPAC Name (6R,7R)-3-(acetyloxymethyl)-7-amino-8-oxo-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylic acid
SMILES CC(=O)OCC1=C(N2C(C(C2=O)N)SC1)C(=O)O

Specifications

Quantity 5 g
Formula Weight 272.28
Percent Purity ≥97.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 7-Aminocephalosporanic Acid