missing translation for 'onlineSavingsMsg'
Learn More

Molecular Probes™ 7-Amino-4-Methylcoumarin, reference standard

Catalog No. A191A
Encompass
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A191A 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A191A Supplier Molecular Probes™ Supplier No. A191

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

7-Amino-4-methylcoumarin (AMC) can be used as a fluorescence reference standard for AMC-based enzyme substrates including AMC-based caspase substrates. | 175.2 g/mol

7-Amino-4-methylcoumarin (AMC) can be used as a fluorescence reference standard for AMC-based enzyme substrates including AMC-based caspase substrates. The fluorescence spectra of AMC is not subject to variability due to pH-dependent protonation/deprotonation when assayed near or above physiological pH.

To make a concentrated stock solution of up to 10 mg/mL, dissolve in either acetone, high quality anhydrous DMSO or DMF. Store frozen, desiccated and protected from light.

Specifications

Quantity 100 mg
Product Type 7-Amino-4-Methylcoumarin
Molecular Weight (g/mol) 175.2
Content And Storage Store at room temperature and protect from light.
Shipping Condition Room Temperature
Label or Dye Methylcoumarin
Label Type Coumarin Dyes
Emission 430 nm
Excitation 351 nm
What solvent should I use to reconstitute 7-Amino-4-Methylcoumarin, reference standard (Cat. No. A191)? Once reconstituted, can the product be aliquoted and stored at -20 degrees C without damaging the molecule?

The 7-Amino-4-Methylcoumarin, reference standard (Cat. No. A191) can be resuspended in DMF or DMSO at a concentration of at least 1 mM. The molecular weight is 175.19 g/mol.
Yes, the solution can be stored in aliquots at -20 degrees C for at least 6 months.

For Research Use Only. Not for use in diagnostic procedures.