Learn More
7-Amino-4-methylcoumarin, 98%
CAS: 26093-31-2 | C10H9NO2 | 175.19 g/mol
Supplier: Thermo Scientific Chemicals A1501706
Description
Used as matrix for analysis of monosulfated disaccharides and as a co-matrix with 6-aza-2-thiothymidine for sulfated neutral and salivated tri-and tetrasaccharides. Reagent for preparing new fluorogenic AMC-based substrates for cystine amino peptidase (and other hydrolases). Also used as reference compound in the enzyme assay.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsUsed as matrix for analysis of monosulfated disaccharides and as a co-matrix with 6-aza-2-thiothymidine for sulfated neutral and salivated tri-and tetrasaccharides. Reagent for preparing new fluorogenic AMC-based substrates for cystine amino peptidase (and other hydrolases). Also used as reference compound in the enzyme assay.
Solubility
Soluble in acetone (10 mg/ml) dimethyl sulfoxide (10 mg/ml) and dimethylformamide.
Notes
Light Sensitive, store in dark. Store in cool, dry place in tightly closed container. Store away from oxidizing agent.
Specifications
| 7-Amino-4-methylcoumarin | |
| 225°C to 228°C | |
| MFCD00006868 | |
| 142231 | |
| Soluble in acetone (10mg/ml) dimethyl sulfoxide (10mg/ml) and dimethylformamide. | |
| CC1=CC(=O)OC2=CC(N)=CC=C12 | |
| 175.19 | |
| CHEBI:51771 | |
| 98% |
| 26093-31-2 | |
| C10H9NO2 | |
| 5 g | |
| 7-amino-4-methylcoumarin, coumarin 120, 7-amino-4-methyl-2h-chromen-2-one, 2h-1-benzopyran-2-one, 7-amino-4-methyl, coumarin, 7-amino-4-methyl, 4-methyl-7-aminocoumarin, 7-amino-4-methyl-chromen-2-one, unii-ocy3jct44x, ccris 4961, ocy3jct44x | |
| GLNDAGDHSLMOKX-UHFFFAOYSA-N | |
| 7-amino-4-methylchromen-2-one | |
| 92249 | |
| 175.19 |
Chemical Identifiers
| 26093-31-2 | |
| 175.19 | |
| GLNDAGDHSLMOKX-UHFFFAOYSA-N | |
| 92249 | |
| 7-amino-4-methylchromen-2-one |
| C10H9NO2 | |
| MFCD00006868 | |
| 7-amino-4-methylcoumarin, coumarin 120, 7-amino-4-methyl-2h-chromen-2-one, 2h-1-benzopyran-2-one, 7-amino-4-methyl, coumarin, 7-amino-4-methyl, 4-methyl-7-aminocoumarin, 7-amino-4-methyl-chromen-2-one, unii-ocy3jct44x, ccris 4961, ocy3jct44x | |
| CHEBI:51771 | |
| CC1=CC(=O)OC2=CC(N)=CC=C12 |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335-H500
EINECSNumber : 247-454-8
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only