missing translation for 'onlineSavingsMsg'
Learn More

7-Amino-4-methylcoumarin, 98%

Catalog No. p-7022306
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA15017MD 250 mg
AAA1501703 1 g
AAA1501706 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA15017MD Supplier Thermo Scientific Chemicals Supplier No. A15017MD
Only null left

CAS: 26093-31-2 | C10H9NO2 | 175.19 g/mol

Used as matrix for analysis of monosulfated disaccharides and as a co-matrix with 6-aza-2-thiothymidine for sulfated neutral and salivated tri-and tetrasaccharides. Reagent for preparing new fluorogenic AMC-based substrates for cystine amino peptidase (and other hydrolases). Also used as reference compound in the enzyme assay.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Used as matrix for analysis of monosulfated disaccharides and as a co-matrix with 6-aza-2-thiothymidine for sulfated neutral and salivated tri-and tetrasaccharides. Reagent for preparing new fluorogenic AMC-based substrates for cystine amino peptidase (and other hydrolases). Also used as reference compound in the enzyme assay.

Solubility
Soluble in acetone (10 mg/ml) dimethyl sulfoxide (10 mg/ml) and dimethylformamide.

Notes
Light Sensitive, store in dark. Store in cool, dry place in tightly closed container. Store away from oxidizing agent.

Chemical Identifiers

CAS 26093-31-2
Molecular Formula C10H9NO2
Molecular Weight (g/mol) 175.19
MDL Number MFCD00006868
InChI Key GLNDAGDHSLMOKX-UHFFFAOYSA-N
Synonym 7-amino-4-methylcoumarin, coumarin 120, 7-amino-4-methyl-2h-chromen-2-one, 2h-1-benzopyran-2-one, 7-amino-4-methyl, coumarin, 7-amino-4-methyl, 4-methyl-7-aminocoumarin, 7-amino-4-methyl-chromen-2-one, unii-ocy3jct44x, ccris 4961, ocy3jct44x
PubChem CID 92249
ChEBI CHEBI:51771
IUPAC Name 7-amino-4-methylchromen-2-one
SMILES CC1=CC(=O)OC2=CC(N)=CC=C12

Specifications

Melting Point 225°C to 228°C
Quantity 250 mg
Beilstein 142231
Solubility Information Soluble in acetone (10mg/ml) dimethyl sulfoxide (10mg/ml) and dimethylformamide.
Formula Weight 175.19
Percent Purity 98%
Chemical Name or Material 7-Amino-4-methylcoumarin
Hazard Category H315-H319-H335-H500
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 247-454-8
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only