missing translation for 'onlineSavingsMsg'
Learn More

7-Amino-4-methylcoumarin, 98%

CAS: 26093-31-2 | C10H9NO2 | 175.19 g/mol

Supplier:  Thermo Scientific Chemicals A1501703

 View more versions of this product

Catalog No. AAA1501703

Only null left
Explore available promotions

Description

Description

Used as matrix for analysis of monosulfated disaccharides and as a co-matrix with 6-aza-2-thiothymidine for sulfated neutral and salivated tri-and tetrasaccharides. Reagent for preparing new fluorogenic AMC-based substrates for cystine amino peptidase (and other hydrolases). Also used as reference compound in the enzyme assay.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Used as matrix for analysis of monosulfated disaccharides and as a co-matrix with 6-aza-2-thiothymidine for sulfated neutral and salivated tri-and tetrasaccharides. Reagent for preparing new fluorogenic AMC-based substrates for cystine amino peptidase (and other hydrolases). Also used as reference compound in the enzyme assay.

Solubility
Soluble in acetone (10 mg/ml) dimethyl sulfoxide (10 mg/ml) and dimethylformamide.

Notes
Light Sensitive, store in dark. Store in cool, dry place in tightly closed container. Store away from oxidizing agent.
Specifications

Specifications

7-Amino-4-methylcoumarin
225°C to 228°C
MFCD00006868
142231
Soluble in acetone (10mg/ml) dimethyl sulfoxide (10mg/ml) and dimethylformamide.
CC1=CC(=O)OC2=CC(N)=CC=C12
175.19
CHEBI:51771
98%
26093-31-2
C10H9NO2
1 g
7-amino-4-methylcoumarin, coumarin 120, 7-amino-4-methyl-2h-chromen-2-one, 2h-1-benzopyran-2-one, 7-amino-4-methyl, coumarin, 7-amino-4-methyl, 4-methyl-7-aminocoumarin, 7-amino-4-methyl-chromen-2-one, unii-ocy3jct44x, ccris 4961, ocy3jct44x
GLNDAGDHSLMOKX-UHFFFAOYSA-N
7-amino-4-methylchromen-2-one
92249
175.19

Chemical Identifiers

26093-31-2
175.19
GLNDAGDHSLMOKX-UHFFFAOYSA-N
92249
7-amino-4-methylchromen-2-one
C10H9NO2
MFCD00006868
7-amino-4-methylcoumarin, coumarin 120, 7-amino-4-methyl-2h-chromen-2-one, 2h-1-benzopyran-2-one, 7-amino-4-methyl, coumarin, 7-amino-4-methyl, 4-methyl-7-aminocoumarin, 7-amino-4-methyl-chromen-2-one, unii-ocy3jct44x, ccris 4961, ocy3jct44x
CHEBI:51771
CC1=CC(=O)OC2=CC(N)=CC=C12
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335-H500

EINECSNumber : 247-454-8

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only