missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ eBioscience™ 7-AAD Viability Staining Solution
Description
Fluorescence is detected in the far red range of the spectrum (650nm long-pass filter)
Specifications
Specifications
| Color | Red |
| Detection Method | Fluorescence |
| For Use With (Application) | Viability Assay |
| For Use With (Equipment) | Flow Cytometer |
| Product Type | Viability Staining Solution |
| Dye Type | Cell-Impermeant |
| Product Line | eBioscience |
| Quantity | 2 mL |
| Emission Wavelength | 535/617 |
For Research Use Only. Not for use in diagnostic procedures.