missing translation for 'onlineSavingsMsg'
Learn More

7,8-Dimethoxy-1H-benzo[d]azepin-2(3H)-one 98.0+%, TCI America™

Catalog No. D48245G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D48245G 5 g
D48241G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D48245G Supplier TCI America Supplier No. D48245G

Chemical Identifiers

CAS 73942-87-7
Molecular Formula C12H13NO3
Molecular Weight (g/mol) 219.24
MDL Number MFCD02091565
InChI Key CPNZASIAJKSBBH-UHFFFAOYSA-N
Synonym 1,3-Dihydro-7,8-dimethoxy-2H-3-benzazepin-2-one
PubChem CID 1482373
IUPAC Name 7,8-dimethoxy-1,3-dihydro-3-benzazepin-2-one
SMILES COC1=C(C=C2C=CNC(=O)CC2=C1)OC

Specifications

Melting Point 246°C
Color White-Yellow
Quantity 5 g
Formula Weight 219.24
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 7,8-Dimethoxy-1H-benzo[d]azepin-2(3H)-one