accessibility menu, dialog, popup

UserName

CH3ONHOOH3C

CAS RN 73942-87-7

IUPAC Name: 7,8-dimethoxy-2,3-dihydro-1H-3-benzazepin-2-one
Synonyms: 7,8-dimethoxy-1,3-dihydro-3-benzazepin-2-one
Molecular Weight (g/mol): 219.24
Molecular Formula: C12H13NO3
InChi Key: CPNZASIAJKSBBH-UHFFFAOYSA-N
SMILES: COC1=C(OC)C=C2C=CNC(=O)CC2=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results