missing translation for 'onlineSavingsMsg'
Learn More

7,8-Dihydroxyflavone Hydrate 98.0+%, TCI America™

Catalog No. D19161G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D19161G 1 g
D19165G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D19161G Supplier TCI America Supplier No. D19161G

Chemical Identifiers

CAS 38183-03-8
Molecular Formula C15H10O4
Molecular Weight (g/mol) 254.24
MDL Number MFCD00006836
InChI Key COCYGNDCWFKTMF-UHFFFAOYSA-N
Synonym 7,8-dihydroxyflavone, 7,8-dihydroxy-2-phenyl-4h-chromen-4-one, 7,8-dihydroxy-flavone, 4h-1-benzopyran-4-one, 7,8-dihydroxy-2-phenyl, 7,8-dhf, 7,8-dihydroxy-2-phenyl-4-benzopyrone, 7,8-dihydroxy-2-phenyl-chromen-4-one, 7,8-dihydroxyflavone hydrate, chembl75267, 7,8-dihydroxy-2-phenyl-4h-1-benzopyran-4-one
PubChem CID 1880
IUPAC Name 7,8-dihydroxy-2-phenyl-4H-chromen-4-one
SMILES OC1=CC=C2C(=O)C=C(OC2=C1O)C1=CC=CC=C1

Specifications

Melting Point 245°C
Quantity 1 g
Formula Weight 254.24
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 7,8-Dihydroxyflavone Hydrate
RTECSNumber DJ3004000
TSCA No