missing translation for 'onlineSavingsMsg'
Learn More

7,8-Dihydroxyflavone hydrate, 97%

Catalog No. p-7030290
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH2785203 1 g
AAH2785206 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH2785203 Supplier Thermo Scientific Chemicals Supplier No. H2785203
Only null left

CAS: 38183-03-8 | C15H10O4 | 254.24 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 38183-03-8
Molecular Formula C15H10O4
Molecular Weight (g/mol) 254.24
MDL Number MFCD00006836
InChI Key COCYGNDCWFKTMF-UHFFFAOYSA-N
Synonym 7,8-dihydroxyflavone, 7,8-dihydroxy-2-phenyl-4h-chromen-4-one, 7,8-dihydroxy-flavone, 4h-1-benzopyran-4-one, 7,8-dihydroxy-2-phenyl, 7,8-dhf, 7,8-dihydroxy-2-phenyl-4-benzopyrone, 7,8-dihydroxy-2-phenyl-chromen-4-one, 7,8-dihydroxyflavone hydrate, chembl75267, 7,8-dihydroxy-2-phenyl-4h-1-benzopyran-4-one
PubChem CID 1880
IUPAC Name 7,8-dihydroxy-2-phenylchromen-4-one
SMILES OC1=CC=C2C(=O)C=C(OC2=C1O)C1=CC=CC=C1

Specifications

Melting Point 250°C to 252°C
Quantity 1 g
Formula Weight 254.24 (Anhydrous)
Percent Purity 97%
Chemical Name or Material 7,8-Dihydroxyflavone hydrate
EINECSNumber 253-812-4
RTECSNumber DJ3004000
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only