missing translation for 'onlineSavingsMsg'
Learn More

7,7-Dimethyl-7H-benzo[c]fluorene 94.0+%, TCI America™

Catalog No. p-7128662
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D51251G 1 g
D51255G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D51251G Supplier TCI America Supplier No. D51251G
Only null left

Chemical Identifiers

CAS 112486-09-6
Molecular Formula C19H16
Molecular Weight (g/mol) 244.337
InChI Key SJVOYVUTRSZMKW-UHFFFAOYSA-N
PubChem CID 59513021
IUPAC Name 7,7-dimethylbenzo[c]fluorene
SMILES CC1(C2=C(C3=CC=CC=C3C=C2)C4=CC=CC=C41)C

Specifications

Melting Point 84°C
Quantity 1 g
Formula Weight 244.34
Percent Purity ≥94.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 7,7-Dimethyl-7H-benzo[c]fluorene