missing translation for 'onlineSavingsMsg'
Learn More
Learn More
7-(2-Chloroethyl)theophylline 98.0+%, TCI America™
Supplier: TCI America C274825G
Specifications
| 7-(2-Chloroethyl)theophylline | |
| 125°C | |
| C9H11ClN4O2 | |
| 25 g | |
| QCIARNIKNKKHFH-UHFFFAOYSA-N | |
| 7-(2-chloroethyl)-1,3-dimethylpurine-2,6-dione | |
| 1882 | |
| ≥98.0% (HPLC,T) |
| 5878-61-5 | |
| White | |
| MFCD00005760 | |
| 7-2-chloroethyl theophylline, benaphyllin, eupnophile, 7-chloroethyl theophylline, theophylline, 7-2-chloroethyl, beta-chloroethyltheophylline, unii-y7202un6b7, 7-beta-chlorethyl theophylline, 7-chloroethyltheophylline, 1h-purine-2,6-dione, 7-2-chloroethyl-3,7-dihydro-1,3-dimethyl | |
| CN1C2=C(C(=O)N(C1=O)C)N(C=N2)CCCl | |
| 242.663 | |
| 242.66 | |
| Crystalline Powder |
Chemical Identifiers
| 5878-61-5 | |
| 242.663 | |
| QCIARNIKNKKHFH-UHFFFAOYSA-N | |
| 1882 | |
| CN1C2=C(C(=O)N(C1=O)C)N(C=N2)CCCl |
| C9H11ClN4O2 | |
| MFCD00005760 | |
| 7-2-chloroethyl theophylline, benaphyllin, eupnophile, 7-chloroethyl theophylline, theophylline, 7-2-chloroethyl, beta-chloroethyltheophylline, unii-y7202un6b7, 7-beta-chlorethyl theophylline, 7-chloroethyltheophylline, 1h-purine-2,6-dione, 7-2-chloroethyl-3,7-dihydro-1,3-dimethyl | |
| 7-(2-chloroethyl)-1,3-dimethylpurine-2,6-dione |
Safety and Handling
RTECSNumber : XH5090000
TSCA : No