accessibility menu, dialog, popup

UserName

CH3NONCH3NNClO

CAS RN 5878-61-5

IUPAC Name: 7-(2-chloroethyl)-1,3-dimethyl-2,3,6,7-tetrahydro-1H-purine-2,6-dione
Synonyms: 7-(2-chloroethyl)theophylline
Molecular Weight (g/mol): 242.66
Molecular Formula: C9H11ClN4O2
InChi Key: QCIARNIKNKKHFH-UHFFFAOYSA-N
SMILES: CN1C2=C(N(CCCl)C=N2)C(=O)N(C)C1=O
Percent Purity 
  • (2)
  • (1)
Physical Form 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)