missing translation for 'onlineSavingsMsg'
Learn More

7-(2-Chloroethyl)theophylline, 97%, Thermo Scientific Chemicals

Catalog No. p-7043022
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ6511103 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAJ6511103 Supplier Thermo Scientific Chemicals Supplier No. J6511103

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

An adenosine receptor antagonist

An adenosine receptor antagonist

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 5878-61-5
Molecular Formula C9H11ClN4O2
Molecular Weight (g/mol) 242.663
MDL Number MFCD00005760
InChI Key QCIARNIKNKKHFH-UHFFFAOYSA-N
Synonym 7-2-chloroethyl theophylline, benaphyllin, eupnophile, 7-chloroethyl theophylline, theophylline, 7-2-chloroethyl, beta-chloroethyltheophylline, unii-y7202un6b7, 7-beta-chlorethyl theophylline, 7-chloroethyltheophylline, 1h-purine-2,6-dione, 7-2-chloroethyl-3,7-dihydro-1,3-dimethyl
PubChem CID 1882
IUPAC Name 7-(2-chloroethyl)-1,3-dimethylpurine-2,6-dione
SMILES CN1C2=C(C(=O)N(C1=O)C)N(C=N2)CCCl

Specifications

Melting Point 122°C to 130°C
Color White
Quantity 1 g
Solubility Information Soluble in DMSO; Moderately soluble in ethanol.
Formula Weight 242.66
Percent Purity 97%
Physical Form Powder
Chemical Name or Material 7-(2-Chloroethyl)theophylline
Hazard Category H302
Hazard Statement GHS H Statement
H302
Harmful if swallowed.
Precautionary Statement P264b-P270-P301+P312-P330-P501c
EINECSNumber 227-553-2
RTECSNumber XH5090000
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only