missing translation for 'onlineSavingsMsg'
Learn More

7,12-Dimethylbenz[a]anthracene 98.0+%, TCI America™

Catalog No. p-7126365
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D06771G 1 g
D06775G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D06771G Supplier TCI America Supplier No. D06771G

Chemical Identifiers

CAS 57-97-6
Molecular Formula C20H16
Molecular Weight (g/mol) 256.348
MDL Number MFCD00003600
InChI Key ARSRBNBHOADGJU-UHFFFAOYSA-N
Synonym dmba, 7,12-dimethylbenz a anthracene, 7,12-dimethylbenzanthracene, 9,10-dimethyl-1,2-benzanthracene, 7,12-dmba, 7,12-dimethylbenzo a anthracene, 7,12-dimethylbenzanthrancene, 6,7-dimethyl-1,2-benzanthracene, 1,4-dimethyl-2,3-benzphenanthrene
PubChem CID 6001
ChEBI CHEBI:254496
IUPAC Name 7,12-dimethylbenzo[a]anthracene
SMILES CC1=C2C=CC3=CC=CC=C3C2=C(C4=CC=CC=C14)C

Specifications

Melting Point 124°C
Color Yellow
Quantity 1 g
Formula Weight 256.35
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 7,12-Dimethylbenz[a]anthracene
RTECSNumber CW3850000
TSCA Yes