missing translation for 'onlineSavingsMsg'
Learn More

7,12-Dimethylbenz[a]anthracene, 98%

Catalog No. AC408181000
Change view
Click to view available options
Quantity:
100 mg
Packaging:
Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC408181000 100 mg Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC408181000 Supplier Thermo Scientific Chemicals Supplier No. 408181000
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 57-97-6 | C20H16 | 256.34 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 57-97-6
Molecular Formula C20H16
Molecular Weight (g/mol) 256.34
MDL Number MFCD00003600
InChI Key ARSRBNBHOADGJU-UHFFFAOYSA-N
Synonym dmba, 7,12-dimethylbenz a anthracene, 7,12-dimethylbenzanthracene, 9,10-dimethyl-1,2-benzanthracene, 7,12-dmba, 7,12-dimethylbenzo a anthracene, 7,12-dimethylbenzanthrancene, 6,7-dimethyl-1,2-benzanthracene, 1,4-dimethyl-2,3-benzphenanthrene
PubChem CID 6001
ChEBI CHEBI:254496
IUPAC Name 7,12-dimethylbenzo[a]anthracene
SMILES CC1=C2C=CC3=CC=CC=C3C2=C(C4=CC=CC=C14)C

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 122°C to 125°C
Color Green to Yellow
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Packaging Glass bottle
Quantity 100 mg
Beilstein 05, III, 2413
Merck Index 15, 3257
Formula Weight 256.34
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material 7, 12-Dimethylbenz[a]anthracene, 99%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Harmful if swallowed.
May cause cancer.
Precautionary Statement GHS P Statement:
Obtain special instructions before use.
IF exposed or concerned: Get medical advice/attention.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 200-359-5
RTECSNumber CW3850000
TSCA TSCA

RUO – Research Use Only