missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HydroSpec™ Solvent K, Ricca Chemical
Specifications
Specifications
| Chemical Name or Material | Solvent KT |
| CAS | 107-98-2, 7446-09-5, 7446-09-5, 67-66-3, 107-98-2 |
| CAS Min % | 5 |
| CAS Max % | 10, 30 |
| Physical Form | Liquid |
| Quantity | 1 L |
| Packaging | Amber Glass Bottle |
| UN Number | UN3092 |
| Specific Gravity | 1.12 |
| Grade | Karl Fischer |