missing translation for 'onlineSavingsMsg'
Learn More

6-(Trifluoromethyl)imidazo[1,2-a]pyridine-2-carboxylic acid hydrate, 95%, Thermo Scientific™

Catalog No. AAH35639MD
Change view
Click to view available options
Quantity:
250mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH35639MD 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH35639MD Supplier Thermo Scientific Chemicals Supplier No. H35639MD

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Chemical Identifiers

CAS 1858251-06-5
Molecular Formula C9H7F3N2O3
Molecular Weight (g/mol) 248.161
MDL Number MFCD22682819
InChI Key HECZGTGLQVJBSB-UHFFFAOYSA-N
Synonym 6-trifluoromethyl imidazo 1,2-a pyridine-2-carboxylic acid hydrate
PubChem CID 112758303
IUPAC Name 6-(trifluoromethyl)imidazo[1,2-a]pyridine-2-carboxylic acid;hydrate
SMILES C1=CC2=NC(=CN2C=C1C(F)(F)F)C(=O)O.O

Specifications

Quantity 250mg
Formula Weight 230.15 (Anhydrous)
Percent Purity 95%
Chemical Name or Material 6-(Trifluoromethyl)imidazo[1,2-a]pyridine-2-carboxylic acid hydrate
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures