accessibility menu, dialog, popup

UserName

NNFFFOHOH2O

CAS RN 1858251-06-5

IUPAC Name: 6-(trifluoromethyl)imidazo[1,2-a]pyridine-2-carboxylic acid hydrate
Synonyms: 6-(trifluoromethyl)imidazo[1,2-a]pyridine-2-carboxylic acid hydrate
Molecular Weight (g/mol): 248.16
Molecular Formula: C9H7F3N2O3
InChi Key: HECZGTGLQVJBSB-UHFFFAOYSA-N
SMILES: O.OC(=O)C1=CN2C=C(C=CC2=N1)C(F)(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)