missing translation for 'onlineSavingsMsg'
Learn More

[6-(Tetrahydropyran-4-yloxy)pyrid-3-yl]methanol, 97%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals CC58309DA

 View more versions of this product

Catalog No. CC58309DA

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

Specifications

Specifications

[6-(Tetrahydropyran-4-yloxy)pyrid-3-yl]methanol
96.5
97%
C11H15NO3
1g
UTEOAQJLPJOMJS-UHFFFAOYSA-N
[6-(oxan-4-yloxy)pyridin-3-yl]methanol
24229579
97%
906352-79-2
100.0
Amber Glass Bottle
MFCD09817494
6-oxan-4-yloxy pyridin-3-yl methanol, 6-tetrahydropyran-4-yloxy pyrid-3-yl methanol, 6-tetrahydropyran-4-yloxy pyridin-3-yl methanol, 6-tetrahydro-2h-pyran-4-yloxy pyridin-3-yl methanol, 6-oxan-4-yl oxy pyridin-3-yl methanol, 5-hydroxymethyl-2-tetrahydropyran-4-yloxy pyridine, 3-pyridinemethanol,6-tetrahydro-2h-pyran-4-yl oxy, 6-tetrahydro-2h-pyran-4-yl oxy pyridin-3-yl methanol, 6-2h-3,4,5,6-tetrahydropyran-4-yloxy-3-pyridyl methan-1-ol
C1COCCC1OC2=NC=C(C=C2)CO
209.245
209.24

Chemical Identifiers

906352-79-2
209.245
UTEOAQJLPJOMJS-UHFFFAOYSA-N
24229579
C1COCCC1OC2=NC=C(C=C2)CO
C11H15NO3
MFCD09817494
6-oxan-4-yloxy pyridin-3-yl methanol, 6-tetrahydropyran-4-yloxy pyrid-3-yl methanol, 6-tetrahydropyran-4-yloxy pyridin-3-yl methanol, 6-tetrahydro-2h-pyran-4-yloxy pyridin-3-yl methanol, 6-oxan-4-yl oxy pyridin-3-yl methanol, 5-hydroxymethyl-2-tetrahydropyran-4-yloxy pyridine, 3-pyridinemethanol,6-tetrahydro-2h-pyran-4-yl oxy, 6-tetrahydro-2h-pyran-4-yl oxy pyridin-3-yl methanol, 6-2h-3,4,5,6-tetrahydropyran-4-yloxy-3-pyridyl methan-1-ol
[6-(oxan-4-yloxy)pyridin-3-yl]methanol
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement:
May cause respiratory irritation.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
Harmful if inhaled.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

SDS Documents

Documents

Promotions

Promotions