missing translation for 'onlineSavingsMsg'
Learn More

[6-(Tetrahydropyran-4-yloxy)pyrid-3-yl]methanol, 97%, Thermo Scientific™

Catalog No. p-3830159
Change view
Click to view available options
Quantity:
250mg
1g
Packaging:
Amber Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
CC58309CB 250mg Amber Glass Bottle
CC58309DA 1g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. CC58309CB Supplier Thermo Scientific Chemicals Supplier No. CC58309CB

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 906352-79-2
Molecular Formula C11H15NO3
Molecular Weight (g/mol) 209.245
MDL Number MFCD09817494
InChI Key UTEOAQJLPJOMJS-UHFFFAOYSA-N
Synonym 6-oxan-4-yloxy pyridin-3-yl methanol, 6-tetrahydropyran-4-yloxy pyrid-3-yl methanol, 6-tetrahydropyran-4-yloxy pyridin-3-yl methanol, 6-tetrahydro-2h-pyran-4-yloxy pyridin-3-yl methanol, 6-oxan-4-yl oxy pyridin-3-yl methanol, 5-hydroxymethyl-2-tetrahydropyran-4-yloxy pyridine, 3-pyridinemethanol,6-tetrahydro-2h-pyran-4-yl oxy, 6-tetrahydro-2h-pyran-4-yl oxy pyridin-3-yl methanol, 6-2h-3,4,5,6-tetrahydropyran-4-yloxy-3-pyridyl methan-1-ol
PubChem CID 24229579
IUPAC Name [6-(oxan-4-yloxy)pyridin-3-yl]methanol
SMILES C1COCCC1OC2=NC=C(C=C2)CO

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250mg
Formula Weight 209.24
Percent Purity 97%
Chemical Name or Material [6-(Tetrahydropyran-4-yloxy)pyrid-3-yl]methanol
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
May cause respiratory irritation.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
Harmful if inhaled.
Precautionary Statement GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.