missing translation for 'onlineSavingsMsg'
Learn More
Learn More
6-tert-Butyl-o-cresol 99.0+%, TCI America™
$76.43 - $367.22
Chemical Identifiers
| CAS | 2219-82-1 |
|---|---|
| Molecular Formula | C11H16O |
| Molecular Weight (g/mol) | 164.25 |
| MDL Number | MFCD00002239 |
| InChI Key | BKZXZGWHTRCFPX-UHFFFAOYSA-N |
| Synonym | 2-tert-Butyl-6-methylphenol |
| PubChem CID | 16678 |
| IUPAC Name | 2-tert-butyl-6-methylphenol |
| SMILES | CC1=CC=CC(=C1O)C(C)(C)C |
Chemical Identifiers
| 2219-82-1 | |
| 164.25 | |
| BKZXZGWHTRCFPX-UHFFFAOYSA-N | |
| 16678 | |
| CC1=CC=CC(=C1O)C(C)(C)C |
| C11H16O | |
| MFCD00002239 | |
| 2-tert-Butyl-6-methylphenol | |
| 2-tert-butyl-6-methylphenol |
Specifications
| 2219-82-1 | |
| MFCD00002239 | |
| 2430 | |
| BKZXZGWHTRCFPX-UHFFFAOYSA-N | |
| 2-tert-butyl-6-methylphenol | |
| 16678 | |
| ≥99.0% (GC) | |
| 6-tert-Butyl-o-cresol |
| C11H16O | |
| 25 g | |
| 2-tert-Butyl-6-methylphenol | |
| CC1=CC=CC(=C1O)C(C)(C)C | |
| 164.25 | |
| 164.25 | |
| Crystalline Lumps |
Safety and Handling
EINECSNumber : (3)-0521
RTECSNumber : SK1578200
TSCA : Yes