missing translation for 'onlineSavingsMsg'
Learn More
Learn More
6-tert-Butyl-o-cresol 99.0+%, TCI America™
Specifications
Specifications
| Quantity | 500 g |
| UN Number | 2430 |
| Formula Weight | 164.25 |
| Percent Purity | ≥99.0% (GC) |
| Physical Form | Crystalline Lumps |
| Chemical Name or Material | 6-tert-Butyl-o-cresol |
Safety and Handling
| EINECSNumber | (3)-0521 |
| RTECSNumber | SK1578200 |
| TSCA | Yes |